missing translation for 'onlineSavingsMsg'
Learn More

SIGNR4/CD209e Antibody (RM0327-9B29), PE, Novus Biologicals™

Product Code. 18416241 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416241 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416241 Supplier Novus Biologicals Supplier No. NBP212431PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

SIGNR4/CD209e Monoclonal antibody specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGNR4/CD209e
Applications Western Blot
Classification Monoclonal
Clone RM0327-9B29
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias mSIGNR4
Host Species Rat
Immunogen Mouse recombinant SIGNR4/CD209e
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.