missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGNR4/CD209e Antibody (RM0327-9B29), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
SIGNR4/CD209e Monoclonal antibody specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SIGNR4/CD209e |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | RM0327-9B29 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | mSIGNR4 |
| Host Species | Rat |
| Immunogen | Mouse recombinant SIGNR4/CD209e |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?