missing translation for 'onlineSavingsMsg'
Learn More

SIGNR1/CD209b Antibody (258829) [DyLight 594], Novus Biologicals™

Product Code. 30737611 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737611 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737611 Supplier Novus Biologicals Supplier No. FAB1836M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

SIGNR1/CD209b Monoclonal antibody specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGNR1/CD209b
Applications Western Blot
Classification Monoclonal
Clone 258829
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1/CD209b, Ser76-Gly325, Accession # Q8CJ91
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170786
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.