missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGNR1/CD209b Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90607CL1
This item is not returnable.
View return policy
Description
SIGNR1/CD209b Monoclonal antibody specifically detects SIGNR1/CD209b in Mouse samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
| SIGNR1/CD209b | |
| Monoclonal | |
| CoraFluor 1 | |
| CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIGNR1/CD209b (Accession#: NP_081248.2; Gln 74-Gly 325). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Sandwich ELISA Detection | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 170786 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction