missing translation for 'onlineSavingsMsg'
Learn More

SIGNR1/CD209b Antibody (001), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18351761 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351761 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351761 Supplier Novus Biologicals Supplier No. NBP290607AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SIGNR1/CD209b Monoclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGNR1/CD209b
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIGNR1/CD209b (NP_081248.2; Gln 74-Gly 325).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170786
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.