Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
Brand:  Epredia™ 8301R
Code : LE
Additional Details : CAS Number : 67-56-1 Weight : 15.00000kg Transport : UN number : 1987 Chem class : 3 Pack group : II
 View more versions of this product
Product Code. 12638106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
  • Excellent for lipid extraction allowing for increased paraffin infiltration
  • May be used as a solvent for special stains
Specifications
Specifications
Flex 80 Dehydrating Reagent
1 gal.