Learn More
Epredia™ Signature Series™ Cyto-Stain™
Achieve results similar to conventional Papanicolaou stains with Epredia™ Signature Series Cyto-Stain™, a uniquely formulated cytoplasmic stain
£43.75 - £123.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10435339
|
Epredia™
7501R |
1 gal. |
£123.00
Each |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10465680
|
Epredia™
7511 |
4 pt. |
£43.75
Pack of 4 |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Use for all gynecological and nongynecological procedures
- Reduces staining time
- Cost-effective, as user performs only one counterstain
- Cyto-Stain provides a full range of familiar cytoplasmic colors and control over the staining intensities
- Cyto-Stain G is a modification to produce greener cyanophilic hues in intermediate and basal cells