missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Bluing Reagent

Product Code. 10259988 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 10259988

Brand: Epredia™ 7341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series Bluing Reagent.

  • Buffered to ensure proper alkalinity (pH = 8.0)
  • Specially formulated to prevent tissue separation from the slide
  • Requires only a one-minute water rinse versus conventional bluing reagents which require up-to five minutes

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
TRUSTED_SUSTAINABILITY

Specifications

Product Type Bluing Reagent
Quantity 4 x 1 gal.
pH 8
Form Liquid
compliance-icons compliance-icons
Product Identifier
  • X4 STAIN BLUING REAGENT 4X1GAL . .
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : ProClin 300
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.