Description
- Clever way to change a disposal container without interrupting the inflow.
- LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height.
- When container is changed, the alert can be muted via the new touchpad.
- Includes touchless alarm-resetting fields for up to five containers, up to five switch inputs and up to five switch outputs for peripheral units, and power supply cable with open end and clamp.
Specifications
Specifications
| Dimensions (L x W x H) | 240 x 140 x 2 mm |
| Liner Material | Stainless Steel |
| Product Type | Mounting Bracket |