missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Signal sequence receptor delta Antibody (26G5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42656G
This item is not returnable.
View return policy
Description
Signal sequence receptor delta Monoclonal specifically detects Signal sequence receptor delta in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Signal sequence receptor delta | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SSR4 | |
| Recombinant human SSR4 (24-144aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 26G5 | |
| Western Blot, ELISA | |
| signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6748 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction