missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Signal peptide peptidase-like 2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | Signal peptide peptidase-like 2B |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18219031
|
Novus Biologicals
NBP2-55073 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694717
|
Novus Biologicals
NBP2-55073-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Signal peptide peptidase-like 2B Polyclonal specifically detects Signal peptide peptidase-like 2B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Signal peptide peptidase-like 2B | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 3.4.23, EC 3.4.23.-, IMP-4, IMP4MGC111084, Intramembrane protease 4, KIAA1532signal peptide peptidase-like 2B, Presenilin-like protein 1, PSL1, SNPPL2B, SPPL2b, SPP-like 2B | |
| SPPL2B | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 56928 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YWAGSRDVKKRYMKHKRDDGPEKQEDEAVDV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title