missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Signal Peptide Peptidase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74111
This item is not returnable.
View return policy
Description
Signal Peptide Peptidase Polyclonal specifically detects Signal Peptide Peptidase in Mouse samples. It is validated for Western Blot.
Specifications
| Signal Peptide Peptidase | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| dJ324O17.1, EC 3.4.23.-, H13hIMP1, histocompatibility (minor) 13, IMP-1, IMP1MSTP086, Intramembrane protease 1, minor histocompatibility antigen 13, minor histocompatibility antigen H13, Presenilin-like protein 3, PSENL3, PSL3IMPAS, Signal peptide peptidase, signal peptide peptidase beta, SPPIMPAS-1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 81502 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9D8V0 | |
| HM13 | |
| Synthetic peptides corresponding to the middle region of H13. Immunizing peptide sequence IKLVFPQDLLEKGLEADNFAMLGLGDIVIPGIFIALLLRFDISLKKNTHT. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction