missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma-1 Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577467
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes a receptor protein that interacts with a variety of psychotomimetic drugs, including cocaine and amphetamines. The receptor is believed to play an important role in the cellular functions of various tissues associated with the endocrine, immune, and nervous systems. As indicated by its previous name, opioid receptor sigma 1 (OPRS1), the product of this gene was erroneously thought to function as an opioid receptor; it is now thought to be a non-opioid receptor. Alternative splicing of this gene results in transcript variants encoding distinct isoforms.
Specifications
| Sigma-1 Receptor | |
| Polyclonal | |
| Unconjugated | |
| SIGMAR1 | |
| Sigma-1 Receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10280, 18391, 29336 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.85 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O55242, Q99720, Q9R0C9 | |
| Sigmar1 | |
| Peptide (C)SRGHSGRYWAEISD, corresponding to amino acid residues 113-126 of human Sigma-1 Receptor | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction