missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGLECL12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38557-20ul
This item is not returnable.
View return policy
Description
SIGLECL12 Polyclonal antibody specifically detects SIGLECL12 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SIGLECL12 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ38600, S2V, sialic acid binding Ig-like lectin 12, sialic acid-binding Ig-like lectin 12, Siglec-12, SIGLECL1Sialic acid-binding Ig-like lectin-like 1, SIGLEC-like 1, Siglec-XII, SLGSiglec-L1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-210 of human SIGLECL12 (NP_443729.1).,, Sequence:, EQKDYLLTMQKSVTVQEGLCVSVLCSFSYPQNGWTASDPVHGYWFRAGDHVSRNIPVATNNPARAVQEETRDRFHLLGDPQNKDCTLSIRDTRESDAGTYVFCVERGNMKWNYKYDQLSVNVTASQDLLSRYRLEVPESVTVQEGLCVSVPCSVLYPHYNWTASSPVYGSWFKEGADIPWDIPVATNTPSG | |
| 20 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 89858 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction