missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC8 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519044
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
SIGLEC8 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3-linked sialic acid. and also binds to alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. SIGLEC8 is expressed specifically on eosinophils. The protein contains 1 copy of a cytoplasmic motif that is referred to as the immunoreceptor tyrosine-based inhibitor motif (ITIM). This motif is involved in downmodulation of cellular responses. The phosphorylated ITIM motif binds to the SH2 domain of PTPN6/SHP-1. The SIGLEC8 gene belongs to the immunoglobulin superfamily.
Specifications
| SIGLEC8 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC8 | |
| CDw329; MGC59785; SAF2; SAF-2; sialic acid binding Ig like lectin 8; sialic acid binding Ig-like lectin 8; sialic acid-binding Ig-like lectin 8; Sialoadhesin family member 2; Siglec; SIGLEC8; Siglec-8; SIGLEC8L | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 27181 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9NYZ4 | |
| SIGLEC8 | |
| Synthetic peptide sequence (SYKSQLNYKTKQ) corresponding to the internal amino acids of SIGLEC8 (aa 134-146). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction