missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC7 Monoclonal Antibody (K94028_7G7), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | SIGLEC7 |
|---|---|
| Clone | K94028_7G7 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30202531
|
Invitrogen™
MA556086 |
200 μg |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195165
|
Invitrogen™
MA555706 |
50 μg |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KKSARPAADV GDIGMKDANT IRGSASQGNL TESWADDNPR HHGLAAHSSG EEREIQYAPL SFHKGEPQDL SGQEATNNEY
SIGLEC7 Monoclonal antibody specifically detects SIGLEC7 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| SIGLEC7 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2 | |
| Recombinant human SIGLEC7 (immunogen range is 381-460). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SIGLEC7 |
| K94028_7G7 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y286 | |
| 27036 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title