Learn More
Invitrogen™ SIGLEC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139734
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RVKTRRKKAA QPVQNTDDVN PVMVSGSRGH QHQFQTGIVS DHPAEAGPIS EDEQELHYAV LHFHKVQPQE PKVTDTEYSE IKIHK Highest antigen sequence indentity to the following orthologs: Rat - 32%, Mouse - 35%.
SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
Specifications
| SIGLEC6 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC6 | |
| CD327; CD33 antigen-like 1; CD33L; CD33L1; CD33L2; CDW327; OBBP1; OB-BP1; obesity-binding protein 1; sialic acid binding Ig like lectin 6; sialic acid binding Ig-like lectin 6; sialic acid-binding Ig-like lectin 6; Siglec; SIGLEC6; Siglec-6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43699 | |
| SIGLEC6 | |
| Recombinant protein corresponding to Human SIGLEC6. Recombinant protein control fragment (Product #RP-90056). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.