Learn More
Invitrogen™ SIGLEC5/SIGLEC14 Monoclonal Antibody (1A5), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528138
Description
This antibody recognizes human Siglec-5 (Sialic acid-binding Ig-like lectin 5), otherwise known as CD170, a novel sialic-acid-binding Ig-like lectin, and member of the Ig superfamily, expressed by dendritic cells (DCs), activated macrophages, neutrophils, and cells of the monocyte/myeloid lineage.
SIGLEC5 is a member of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. These cell surface lectins are characterized by structural motifs in the immunoglobulin (Ig)-like domains and sialic acid recognition sites in the first Ig V set domain. The encoded protein is a member of the CD33-related subset of Siglecs and inhibits the activation of several cell types including monocytes, macrophages and neutrophils. Binding of group B Streptococcus (GBS) to the encoded protein plays a role in GBS immune evasion.
Specifications
| SIGLEC5/SIGLEC14 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.09% sodium azide | |
| O15389, Q08ET2 | |
| SIGLEC14, SIGLEC5 | |
| Siglec-5-Fc protein, consisting of the full-length extracellular region of human Siglec-5, fused with the Fc region of human IgG1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 1A5 | |
| FITC | |
| SIGLEC5 | |
| CD170; CD33 antigen-like 2; CD33L2; LLNLR-470E3.1; OB-binding protein 2; OBBP2; OB-BP2; obesity-binding protein 2; sialic acid binding Ig like lectin 14; sialic acid binding Ig like lectin 5; sialic acid binding Ig-like lectin 14; sialic acid binding Ig-like lectin 5; sialic acid-binding Ig-like lectin 14; Sialic acid-binding Ig-like lectin 5; sialic acid-binding immunoglobulin-like lectin 5; SIGLEC14; siglec-14; SIGLEC5; Siglec-5; UNQ294/PRO333 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100049587, 8778 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.