missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114527
This item is not returnable.
View return policy
Description
Positive controls that may be used include: HeLa Cell Lysate, HepG2 Cell Lysate , SK-N-SH Cell Lysate , Human Liver Lysate, Mouse Liver Lysate.
Binds sialylated glycoproteins.
Specifications
| SIGLEC15 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC15 | |
| CD33 antigen-like 3; CD33 molecule-like 3; Cd33l3; EG620235; HsT1361; RGD1565948; sialic acid binding Ig like lectin 15; sialic acid binding Ig-like lectin 15; sialic acid-binding Ig-like lectin 15; Siglec; Siglec 15; SIGLEC15; Siglec-15; SIGLEC-I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284266, 498888, 620235 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.0 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6ZMC9 | |
| SIGLEC15 | |
| Anti-SIGLEC15 antibody was raised against a peptide corresponding to 14 amino acids near the carboxyterminus of human SIGLEC15. The immunogen is located within amino acids 270-320 of SIGLEC15. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction