missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555501
This item is not returnable.
View return policy
Description
Immunogen sequence: DGRFWIRVQE SVMVPEGLCI SVPCSFSYPR QDWTGSTPAY GYWFKAVTET TKGAPVATNH QSREVEMSTR GRFQLTGDPA KGNCSLVIRD AQMQDESQY Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 47%.
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
Specifications
| SIGLEC10 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC10 | |
| PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 89790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LC7 | |
| SIGLEC10 | |
| Recombinant protein corresponding to Human SIGLEC10. Recombinant protein control fragment (Product #RP-90295). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction