Learn More
Invitrogen™ SIGLEC10 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547257
Description
In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) Siglec-5 is observed and less than 1% cross-reactivity with rhSiglec-2, rhSiglec-3, rhSiglec-7, rhSiglec-9, and recombinant mouse Siglec-F is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
Specifications
| SIGLEC10 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC10 | |
| PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 89790 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q96LC7 | |
| SIGLEC10 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-10 Met17-Thr546. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.