Learn More
Siglec-F Rat anti-Mouse, PE, Clone: E50-2440, BD
Rat Monoclonal Antibody
£109.23 - £265.35
Specifications
| Antigen | Siglec-F |
|---|---|
| Clone | E50-2440 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811619
|
BD Biosciences
562068 |
25 μg |
£114.00 £109.23 / Each Save £4.77 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15816768
|
BD Biosciences
552126 |
0.1 mg |
£269.00 £265.35 / Each Save £3.65 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The E50-2440 antibody reacts with Siglec-F, the sixth siglec protein to be reported in the mouse. Siglecs are the sialic acid-binding immunoglobulin superfamily lectins defined in the human, each of which has a distinctive expression pattern in the hematopoietic system and at least some of which are known to mediate cell-cell interactions. Orthologous proteins of human Siglec-1 (Sialoadhesin or CD169), Siglec-2 (CD22), and Siglec-4 (myelin-associated glycoprotein) have been characterized in the mouse. Human Siglec-3 (CD33) and Siglecs-5 through -10 are encoded by a cluster of closely related genes, and each has two cytoplasmic ITIM (Immunoreceptor Tyrosine-based Inhibitory Motifs). Similarly, mouse Siglec-F is encoded by a gene in a syntenic cluster in the mouse, and the protein has sialic acid-binding activity and an intracytoplasmic ITIM. Its expression pattern differs from those of the human Siglec-3-related proteins in that it is found on immature cells of the myelomonocytic lineage, with reduced expression on mature neutrophils and monocytes, and not on lymphoid cells. It has been proposed that mAb E50-2440 may be used for identification of immature myelomonocytic cells in the mouse.
Specifications
| Siglec-F | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse Siglec-F and human IgG Fc recombinant fusion protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| E50-2440 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.