missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-F Rat anti-Mouse, BB515, Clone: E50-2440, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 564514
This item is not returnable.
View return policy
Description
Siglec-F Monoclonal antibody specifically detects Siglec-F in Mouse samples. It is validated for Flow cytometry.
Specifications
| Siglec-F | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| E50-2440 | |
| Brilliant Blue 515 (BB515) | |
| Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5; CD170 | |
| Mouse Siglec-F and human IgG Fc recombinant fusion protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
| IgG2a κ, LOU/M IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction