missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-F Rat anti-Mouse, APC-Cy7, Clone: E50-2440, BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 565527
This item is not returnable.
View return policy
Description
Siglec-F Monoclonal antibody specifically detects Siglec-F in Mouse samples. It is validated for Flow cytometry.
Specifications
| Siglec-F | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| E50-2440 | |
| APC-Cyanine7 | |
| Siglecf; Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5 | |
| Mouse Siglec-F and human IgG Fc recombinant fusion protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with APC-Cy7 under optimum conditions, and unconjugated antibody and free APC-Cy7 were removed. | |
| IgG2a κ, LOU/M IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction