missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-9 Antibody (191240), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1139PEATT594
This item is not returnable.
View return policy
Description
Siglec-9 Monoclonal antibody specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry
Specifications
| Siglec-9 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 191240 | |
| Flow Cytometry | |
| CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-9, Gln18-Gly348 (predicted), Accession # Q9Y336 | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 27180 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction