missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-8 Antibody (837535), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7975L
This item is not returnable.
View return policy
Description
Siglec-8 Monoclonal antibody specifically detects Siglec-8 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Siglec-8 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27181 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 837535 | |
| Flow Cytometry, CyTOF-ready | |
| CD329 antigen, CDw329, MGC59785, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec-8, SIGLEC8Lsialic acid-binding Ig-like lectin 8 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-8, Met17-Ala363, Accession # Q9NYZ4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction