missing translation for 'onlineSavingsMsg'
Learn More

Siglec-5/CD170 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £255.00

Specifications

Antigen Siglec-5/CD170
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655628
View Documents
Novus Biologicals
NBP2-99876-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18620579
View Documents
Novus Biologicals
NBP2-99876-50ul
50 μg
£178.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Siglec-5/CD170 Polyclonal antibody specifically detects Siglec-5/CD170 in Human samples. It is validated for ELISA
Specifications

Specifications

Siglec-5/CD170
ELISA
Unconjugated
Rabbit
Immunology
0.2 um filtered solution in PBS
8778
IgG
Antigen and protein A Affinity-purified
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
Human
CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5
Produced in rabbits immunized with purified, recombinant Human Siglec-5/CD170 (Accession#: O15389; Met1-Thr 434)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.