missing translation for 'onlineSavingsMsg'
Learn More

Siglec-5/CD170 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18324317 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324317 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324317 Supplier Novus Biologicals Supplier No. NBP299876V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Siglec-5/CD170 Polyclonal specifically detects Siglec-5/CD170 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-5/CD170
Applications ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Siglec-5/CD170 (O15389; Met1-Thr 434).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8778
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.