missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-5/CD170 Antibody (07), Novus Biologicals™
Mouse Monoclonal Antibody
£181.00 - £251.00
Specifications
| Antigen | Siglec-5/CD170 |
|---|---|
| Clone | 07 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669777
|
Novus Biologicals
NBP3-06442-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606627
|
Novus Biologicals
NBP3-06442-50ul |
50 μg |
£181.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-5/CD170 Monoclonal antibody specifically detects Siglec-5/CD170 in Human samples. It is validated for Flow CytometrySpecifications
| Siglec-5/CD170 | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Siglec-5/CD170 (Uniprot#: O15389; Met1-Thr 434). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 07 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| 0.2 um filtered solution in PBS | |
| 8778 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title