missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-3/CD33 Antibody (hP67.6 (Gemtuzumab)), Janelia Fluor™ 549, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-75905JF549
This item is not returnable.
View return policy
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications
| Siglec-3/CD33 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD33 | |
| Gemtuzumab was generated by huminization of the murine P67.6 clone, raised against CD33-transfected FMY9S5 cells of human origin. CD33 is a transmembrane receptor expressed on predominantly on cells of the myeloid lineage, but it can also be found on some lymphoid cells. | |
| 0.1 mL | |
| Primary | |
| Recognises human CD33. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunofluorescence | |
| hP67.6 (Gemtuzumab) | |
| Flow Cytometry, Immunofluorescence | |
| CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
| Human | |
| Protein A purified | |
| Immunology | |
| 945.0 | |
| Human | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction