missing translation for 'onlineSavingsMsg'
Learn More

Siglec-3/CD33 Antibody (hP67.6 (Gemtuzumab)), Allophycocyanin, Novus Biologicals™

Product Code. 18366560 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366560 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366560 Supplier Novus Biologicals Supplier No. NBP275905APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-3/CD33
Applications Flow Cytometry
Classification Monoclonal
Clone hP67.6 (Gemtuzumab)
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Gene Symbols CD33
Host Species Human
Immunogen Gemtuzumab was generated by huminization of the murine P67.6 clone, raised against CD33-transfected FMY9S5 cells of human origin. CD33 is a transmembrane receptor expressed on predominantly on cells of the myeloid lineage, but it can also be found on some lymphoid cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 945
Test Specificity Recognises human CD33.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.