missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Siglec-3/CD33 Antibody (Gemtuzumab) - Humanized, IgG4SP, Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Specifications

Antigen Siglec-3/CD33
Clone Gemtuzumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30045678
View Documents
Novus Biologicals™
NBP3-28474-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045788
View Documents
Novus Biologicals™
NBP3-28474-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044724
View Documents
Novus Biologicals™
NBP3-28474-100ug
100 μg
£247.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

Siglec-3/CD33
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Immunology
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Siglec-3/ CD33
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Gemtuzumab
Monoclonal
Purified
RUO
Human
P20138
945
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.