missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Siglec-3/CD33 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | BLR061G |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 20 μg/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Formulation | Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA |
| Gene Alias | CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 |
| Host Species | Rabbit |
| Immunogen | Immunogen was a peptide representing a region between residue 314 and the C-terminus (residue 364) of human Myeloid cell surface antigen CD33 using the numbering given in entry NP_001763.3 (Gene ID 945). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?