missing translation for 'onlineSavingsMsg'
Learn More

Siglec-2/CD22 Antibody (340), DyLight 350, Novus Biologicals™

Product Code. 18354360 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354360 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354360 Supplier Novus Biologicals Supplier No. NBP290030UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-2/CD22
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 340
Conjugate DyLight 350
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM Sodium Borate
Gene Alias B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Siglec-2/CD22 (P20273-1; Met1-Arg687).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 933
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.