Learn More
Siglec-2/CD22 Antibody (219934), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1968A
Product Code. 18698533
  • £516.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Siglec-2/CD22
Monoclonal
APC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Immunocytochemistry, CyTOF
219934
Flow Cytometry, Immunocytochemistry, CyTOF-ready
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14
Mouse myeloma cell line NS0-derived recombinant human Siglec-2/CD22, Asp20-Arg687, Accession # CAA42006
0.1 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology
933
Store at 4°C in the dark.
IgG1