missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (7-239), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37729PECY7
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| SIGLEC1 | |
| Human rhinovirus 14-infected monocyte-derived dendritic cells. | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody 7-239 recognizes CD169 (sialoadhesin, Siglec-1), a 210 kDa type I transmembrane glycoprotein expressed on macrophages and dendritic cells. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 7-239 | |
| Flow Cytometry | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Immunology | |
| 6614.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction