missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGIRR Rabbit anti-Human, Clone: 10, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £458.00
Specifications
| Antigen | SIGIRR |
|---|---|
| Clone | 10 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207707
|
Novus Biologicals
NBP2-90067-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18261906
|
Novus Biologicals
NBP2-90067-100UL |
100 μg |
£458.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIGIRR Monoclonal specifically detects SIGIRR in Human samples. It is validated for Flow Cytometry.Specifications
| SIGIRR | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 59307 | |
| IgG | |
| Protein A purified |
| 10 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8MGC110992, Toll/interleukin-1 receptor 8 | |
| Recombinant Human SIGIRR Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title