missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIGIRR Monoclonal Antibody (161917)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA560915

Product Code. 30536222

  • £448.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

SIGIRR (single Ig IL-1R-related molecule) is a novel member of the interleukin 1 receptor (IL-1R) superfamily. Although it shows the typical conserved motifs that characterize the IL-1R and Toll superfamily, it is structurally and functionally distinct from both. SIGIRR has only one Ig domain in its extracellular portion whereas the IL-1R family contains three Ig folds. An unusually long cytoplasmic domain is reminiscent of the structure of drosophila Toll, yet the SIGIRR peptide sequence is more closely related to IL-1RI. A chimera, in which the SIGIRR intracellular domain was fused to the IL-1R extracellular domain, did not activate NFkappaB unlike similar fusion proteins of other IL-1R related molecules. Phylogenetic data that suggest that the plasma membrane protein SIGIRR is a close functional relative of MyD88 in mammals.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIGIRR
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9JLZ8
SIGIRR
Mouse myeloma cell line NS0-derived recombinant mouse SIGIRR, Met1-His117.
500 μg
Primary
Mouse
Antibody
IgG1
Western Blot
161917
Unconjugated
SIGIRR
AI256711; IL-1R8; MGC110992; Sigirr; single Ig and TIR domain containing; single Ig IL-1 receptor related protein; single Ig IL-1-related receptor; Single Ig IL-1R-related molecule; single Ig IL-1R-related protein; single immunoglobulin and toll-interleukin 1 receptor (TIR) domain; single immunoglobulin domain IL1R1 related; single immunoglobulin domain-containing IL1R-related protein; TIR8; Toll/interleukin-1 receptor 8; UNQ301/PRO342
Rat
Protein A/G
RUO
24058
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.