missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGIRR Antibody (010), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90067DL594
This item is not returnable.
View return policy
Description
SIGIRR Monoclonal specifically detects SIGIRR in Human samples. It is validated for Flow Cytometry.
Specifications
| SIGIRR | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 59307 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 10 | |
| Flow Cytometry | |
| single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8MGC110992, Toll/interleukin-1 receptor 8 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SIGIRR (Q6IA17-1; Met1-His118). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction