Learn More
Endecotts™ Stainless Steel Perforated Plate Sieve, 200mm
Product Code. 15299343
Product Code. 15299343
Brand: Endecotts™ 200SIR4.00
Only null left
Add to basket
Add to basket
For use with all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Perforated Plate Sieve, 200mm includes round holes in a a Stainless Steel plate.
  • Stainless Steel precision frame (ensures consistent nestability)
  • Stainless Steel round hole perforated plate
  • Precise aperture in accordance with ISO 3310-1 / BS410
  • Natural fillet (free flowing of sample)
  • Totally sealed (no crevice to lose material)
  • Evenly tensioned mesh (ensures accurate analysis)
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number (ensures full traceability)