missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Sierra™ Facemasks

Product Code. 15674894 Shop All DuPont Products
Click to view available options
:
Width (Metric): 23 cm
This item is not returnable. View return policy

Product Code. 15674894

Brand: DuPont™ D13396008

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides process protection from human contamination. Dupont™ Sierra™ Facemasks include bound ties and pleated rayon outer facing.

  • Pleated rayon outer facing
  • Bound Tyvek™ ties
  • Metal nose piece
  • 23 cm width
  • White, One size fits most
  • Not clean-processed and not sterilized
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Face Mask
Color White
Material Rayon
Size Standard
Sterility Non-sterile
Certifications/Compliance (Europe) Suitable for use in GMP class C/D cleanrooms
Cleanroom Class Class 1000 (ISO 6)
Disposable Single-use
For Use With (Application) Cleanrooms, Pharmaceutical, Medical Device, Manufacturing, Food Processing
Includes Bound Ties, Metal Nose Piece
Model D13396008
Width (Metric) 23 cm
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.