missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIDT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562591
This item is not returnable.
View return policy
Description
Immunogen sequence: KKAAITVQRK DFPSNSFYVV VVVKTEDQAC GGSLPFYPFA EDEPVDQGHR QKTLSVLVSQ AVTSE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The SID1 transmembrane family member (SIDT2) is a highly glycosylated, multipass transmembrane protein that localizes to the lysosomal membrane. SIDT2 is highly expressed in liver, brain and kidney, with no or little expression in skeletal muscles, heart or other tissues. Little is known of the function of SIDT2.
Specifications
| SIDT2 | |
| Polyclonal | |
| Unconjugated | |
| SIDT2 | |
| SIDT2, SID1 transmembrane family member 2, CGI-40 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBJ9 | |
| SIDT2 | |
| Recombit protein corresponding to Human SIDT2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction