missing translation for 'onlineSavingsMsg'
Learn More

SIDT2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18421186 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421186 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421186 Supplier Novus Biologicals Supplier No. NBP282009JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIDT2 Polyclonal antibody specifically detects SIDT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIDT2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias DKFZp686L17253, FLJ90656, SID1 transmembrane family member 2, SID1 transmembrane family, member 2, UNQ685/PRO1325
Host Species Rabbit
Immunogen Rabbit polyclonal SIDT2 antibody was raised against an 18 amino acid peptide near the amino terminus of human SIDT2. The immunogen is located within the first 50 amino acids of SIDT2. Amino Acid Squence: PKNVSQKDAEFERTYVDE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51092
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.