Learn More
SIDT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55352-1-AP-150UL
 View more versions of this product
Product Code. 16866825
  • £349.00 / 150µL
Description
This antibody is specific to SIDT1.
SIDT1 belongs to the SID1 family.
Specifications
Specifications
SIDT1
Polyclonal
Unconjugated
SIDT1
B830021E24Rik, SID 1, SID1, SIDT1
Rabbit
Antigen Affinity Chromatography
RUO
320007, 54847
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AXF6, Q9NXL6
Sidt1
Peptide
150 μL
Primary
Human, Mouse