missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIDT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55352-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to SIDT1.
SIDT1 belongs to the SID1 family.Specifications
| SIDT1 | |
| Polyclonal | |
| Unconjugated | |
| SIDT1 | |
| B830021E24Rik, SID 1, SID1, SIDT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 320007, 54847 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AXF6, Q9NXL6 | |
| Sidt1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction