missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sibutramine Hydrochloride Monohydrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Sibutramine HCl |
| CAS | 125494-59-9 |
| Analyte or Component Names | Sibutramine HCl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 Cl N . Cl H . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sibutramine Hydrochloride Monohydrate, BTS 54524, Reductil, Meridia, 1-(4-Chlorophenyl)-N,N-dimethyl-alpha-(2-methylpropyl)cyclobutanemethanamine hydrochloride hydrate (1:1:1), 1-(4-Chlorophenyl)-N,N-dimethyl-alpha-(2-methylpropyl)cyclobutanemethanamine hydrochloride monohydrate |
| InChI Formula | InChI=1S/C17H26ClN.ClH.H2O/c1-13(2)12-16(19(3)4)17(10-5-11-17)14-6-8-15(18)9-7-14;;/h6-9,13,16H,5,10-12H2,1-4H3;1H;1H2 |
| Solution Type | Neat |
| SMILES | O.Cl.CC(C)CC(N(C)C)C1(CCC1)c2ccc(Cl)cc2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction