missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIAH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105291
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SIAH2.
SIAH-2 (seven in absentia homolog 2) is an E3 ligase that catalyzes ubiquitination and proteasome-mediated degradation of protein substrates. SIAH-2 encodes a 324 amino acid protein that shares 77% identity with human SIAH-1 and 68% identity with the Drosophila sina (seven in absentia) gene, on which development of the Drosophila R7 photoreceptor is dependent. SIAH-2 targets TRAF2 (which regulates cell responses to stress and cytokines through the regulation of key stress-signaling cascades) for degradation under stress conditions such as hypoxia. It targets HIF-1 alpha prolyl hydroxylase 3 (PHD3) for degradation upon exposure to hypoxic conditions, which coincides with an increase in SIAH-2 transcription. SIAH-2 can decrease TNF-alpha-dependent induction of JNK activity and transcriptional activation of NFkB. SIAH-2 null mice subjected to hypoxia display an impaired respiratory response and reduced levels of hemoglobin.
Specifications
| SIAH2 | |
| Polyclonal | |
| Unconjugated | |
| SIAH2 | |
| AA415433; E3 ubiquitin-protein ligase SIAH2; EC 6.3.2; hSiah2; mSiah2; RING-type E3 ubiquitin transferase SIAH2; seven in absentia 2; seven in absentia homolog 2; siah E3 ubiquitin protein ligase 2; SIAH2; siah-2; Sinh2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140593, 20439, 6478 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43255, Q06986, Q8R4T2 | |
| SIAH2 | |
| A synthesized peptide derived from human SIAH2(Accession O43255), corresponding to amino acid residues F246-C296. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction