missing translation for 'onlineSavingsMsg'
Learn More

SIAH2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-99033C

Product Code. 18579381

  • £322.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SIAH2 Polyclonal antibody specifically detects SIAH2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIAH2
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Western Blot
DyLight 650
E3 ubiquitin-protein ligase SIAH2, EC 6.3.2, EC 6.3.2.-, hSiah2, seven in absentia (Drosophila) homolog 2, Seven in absentia homolog 2, seven in absentia homolog 2 (Drosophila), siah-2
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SIAH2.
0.1 mL
Apoptosis, Cancer, Cell Cycle and Replication
6478
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.