missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAH2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99033AF594
This item is not returnable.
View return policy
Description
SIAH2 Polyclonal antibody specifically detects SIAH2 in Human samples. It is validated for Western Blot
Specifications
| SIAH2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| E3 ubiquitin-protein ligase SIAH2, EC 6.3.2, EC 6.3.2.-, hSiah2, seven in absentia (Drosophila) homolog 2, Seven in absentia homolog 2, seven in absentia homolog 2 (Drosophila), siah-2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SIAH2. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication | |
| 6478 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction