missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIAH1 Polyclonal Antibody
Description
A suggested positive control for this product is human brain lysate.
This gene encodes a protein that is a member of the seven in absentia homolog family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson's disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
Specifications
Specifications
| Antigen | SIAH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | SIAH1 |
| Gene Accession No. | Q8IUQ4 |
| Gene Alias | AA960570; AA982064; AI853500; E3 ubiquitin-protein ligase SIAH1; E3 ubiquitin-protein ligase SIAH1A; E3 ubiquitin-protein ligase SIAH1B; FLJ08065; h HUMSIAH; hSIAH1; HUMSIAH; mSiah-1a; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH1A; RING-type E3 ubiquitin transferase SIAH1B; seven in absentia 1A; seven in absentia 1B; seven in absentia homolog 1; Seven in absentia homolog 1a; Seven in absentia homolog 1b; SIAH; siah E3 ubiquitin protein ligase 1; SIAH1; siah-1; Siah1a; Siah-1a; Siah1b; siah-1b; Sinh1a; Sinh1b |
| Gene Symbols | SIAH1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?